L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR222672500
Price (MYR)
397.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
397.00
Add to cart
Specifications
Chemical Name or MaterialL(+)-Potassium hydrogen tartrate
CAS868-14-4
ColorColorless or White
Density1.9500g/mL
EINECS Number212-769-1