Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass Bottle
Catalog number ACR222165000
Price (MYR)
637.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
637.00
Add to cart
Chemical Identifiers
CAS2486-69-3
IUPAC Name4-amino-3-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeyJNFGLYJROFAOQP-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown to grey
Assay (HPLC)≥94.0%
Melting Point (clear melt)182.0-191.0?C
FormPowder
For UV/VIS spectroscopy
RUO – Research Use Only