Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
CAS: 5329-14-6 | H3NO3S | 97.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic Bottle
Catalog number ACR222070025
Price (MYR)
986.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
986.00
Add to cart
Chemical Identifiers
CAS14914-84-2
IUPAC Nametrichloroholmium hexahydrate
Molecular FormulaCl3H12HoO6
InChI KeyBUQFCXABQYNXLP-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.Cl[Ho](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.