Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
CAS: 5329-14-6 | H3NO3S | 97.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number ACR222070010
Price (MYR)
633.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
633.00
Add to cart
Chemical Identifiers
CAS16373-93-6
IUPAC Name(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H13N5O3
InChI KeyOLXZPDWKRNYJJZ-YUZWJPFSNA-N
SMILESNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)5.0 to 8.0% (theory 6.69%)
Optical Rotation-25.3? ± 1.5? (c = 0.5, Water)
Appearance (Color)White
Assay (HPLC)≥98.5%
Melting Point (clear melt)188.0-194.0?C