Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 65-85-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR221800010
Price (MYR)
1,153.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,153.00
Add to cart
Specifications
Chemical Name or MaterialBenzoic acid
Name NoteFor analysis
CAS65-85-0
Melting Point121.0°C to 123.0°C
Boiling Point249.0°C
View more
For analysis
RUO – Research Use Only