2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 mL
5 mL
Packaging:
Glass bottle
Catalog number ACR221190250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormRod, 3mm (0.1in) diameter (Glassy Carbon type-1, maximimum service temperature 1000?C).
Assay from Supplier's CofA>99.9%