Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR220892500
Price (MYR)
436.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt
Name Notep.a.
pH7.5 to 9.2 (5% soln. at 20°C)
CAS127-09-3
Melting Point324.0°C
View more
Biological buffer
RUO – Research Use Only