Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR220890010
Price (MYR)
929.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
929.00
Add to cart
Chemical Identifiers
CAS64169-34-2
IUPAC Name5-bromo-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC8H5BrO2
InChI KeyIUSPXLCLQIZFHL-UHFFFAOYSA-N
SMILESBrC1=CC=C2C(=O)OCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5% (non-U.S. sourced material)
Water Content (Karl Fischer Titration)≤1.0% (non-U.S. sourced material)
Appearance (Color)White to cream to brown or yellow to orange
CommentMaterial Sourced in the U.S. and in other countries
Melting Point (clear melt)161.0-168.0°C
View more
Biological buffer
RUO – Research Use Only