Gallic acid monohydrate, Thermo Scientific Chemicals
Gallic acid monohydrate, Thermo Scientific Chemicals
Gallic acid monohydrate, Thermo Scientific Chemicals
Gallic acid monohydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallic acid monohydrate, Thermo Scientific Chemicals
CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR220765000
Price (MYR)
1,182.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,182.00
Add to cart
Specifications
Chemical Name or MaterialGallic acid monohydrate
Name Notec.p.
CAS5995-86-8
Melting Point251.0°C
Health Hazard 1Warning