1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number ACR220622500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36070-80-1
IUPAC Name5-chloropyrazine-2-carboxylic acid
Molecular FormulaC5H3ClN2O2
InChI KeyFXJOTWLLDJYKAG-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C(Cl)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Assay (Silylated GC)≥94.0%