1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
250 mL
Packaging:
Glass bottle
Catalog number ACR220620010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS352-11-4
IUPAC Name1-(chloromethyl)-4-fluorobenzene
Molecular FormulaC7H6ClF
InChI KeyIZXWCDITFDNEBY-UHFFFAOYSA-N
SMILESFC1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5120 to 1.5140 (20°C, 589 nm)
Appearance (Form)Liquid