Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR220345000
Price (MYR)
669.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
669.00
Add to cart
Chemical Identifiers
CAS108-59-8
IUPAC Name1,3-dimethyl propanedioate
Molecular FormulaC5H8O4
InChI KeyBEPAFCGSDWSTEL-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
Biological buffer for analysis
RUO – Research Use Only