Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR220341000
Price (MYR)
566.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
566.00
Add to cart
Specifications
Chemical Name or MaterialCitric acid
CAS77-92-9
Melting Point153.0°C
Health Hazard 1Warning
ColorWhite
View more
Biological buffer for analysis
RUO – Research Use Only