Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number ACR220340025
Price (MYR)
1,519.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,519.00
Add to cart
Chemical Identifiers
CAS84468-52-0
IUPAC Name2,3-dimethylpiperazine
Molecular FormulaC6H14N2
InChI KeyCOWPTMLRSANSMQ-UHFFFAOYNA-N
SMILESCC1NCCNC1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Appearance (Color)Pale yellow to yellow
Melting Point (clear melt)81.0-90.0?C
FormCrystals or powder or crystalline powder
Biological buffer for analysis
RUO – Research Use Only