Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number ACR220321000
Price (MYR)
343.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
343.00
Add to cart
Chemical Identifiers
CAS1676-63-7
IUPAC Name1-(4-ethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O2
InChI KeyYJFNFQHMQJCPRG-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point35°C to 38°C
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystals and/or chunks or low melting solid