Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number ACR220320010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with KMnO4>=97.5 %
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder or crystals