sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR220290025
Price (MYR)
1,027.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,027.00
Add to cart
Chemical Identifiers
CAS3069-40-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)> 97.0%