Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR220052500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS582-52-5
IUPAC Name5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular FormulaC12H20O6
InChI KeyKEJGAYKWRDILTF-UHFFFAOYNA-N
SMILESCC1(C)OCC(O1)C1OC2OC(C)(C)OC2C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
GC>=98.0 %
Sulfated ash=<0.2 %
Appearance (Color)White to yellow
Infrared spectrumConforms
View more
For analysis
RUO – Research Use Only