Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR220050050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS5333-86-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
For analysis
RUO – Research Use Only