Acetic acid, ammonium salt, Thermo Scientific Chemicals
Acetic acid, ammonium salt, Thermo Scientific Chemicals
Acetic acid, ammonium salt, Thermo Scientific Chemicals
Acetic acid, ammonium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, ammonium salt, Thermo Scientific Chemicals
CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Plastic bottle
Catalog number ACR219990500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS13414-95-4
IUPAC Name4,5,6,7-tetrahydro-1-benzothiophen-4-one
Molecular FormulaC8H8OS
InChI KeyGJEKNELSXNSYAQ-UHFFFAOYSA-N
SMILESO=C1CCCC2=C1C=CS2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Melting point34°C to 40°C
Appearance (Form)Low melting solid or liquid after melting
GC>=96.0 %
For HPLC
RUO – Research Use Only