Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number ACR219682500
Price (MYR)
588.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
588.00
Add to cart
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=99.0 %
Water=<0.005 %