Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR219680500
Price (MYR)
365.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
365.00
Add to cart
Chemical Identifiers
CAS26386-88-9
IUPAC Name{[azido(phenoxy)phosphoryl]oxy}benzene
Molecular FormulaC12H10N3O3P
InChI KeySORGEQQSQGNZFI-UHFFFAOYSA-N
SMILES[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
Refractive Index1.5485-1.5535 @ 20°C