Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR219221000
Price (MYR)
813.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
813.00
Add to cart
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
FormClear liquid
Viscosity450 - 550 cSt at 25 deg C
Identification (FTIR)Conforms
Appearance (Color)Colorless