1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals
1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals
1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals
1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals
CAS: 627-31-6 | C3H6I2 | 295.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR218600250
Price (MYR)
1,223.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,223.00
Add to cart
Chemical Identifiers
CAS89-61-2
IUPAC Name1,4-dichloro-2-nitrobenzene
Molecular FormulaC6H3Cl2NO2
InChI KeyRZKKOBGFCAHLCZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)52.0-58.0?C
Assay (GC)≥97.5%