Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
CAS: 458-37-7 | C21H20O6 | 368.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR218580500
Price (MYR)
1,946.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,946.00
Add to cart
Chemical Identifiers
CAS90325-47-6
IUPAC Name(2R,3S)-hexane-1,2,3-triol
Molecular FormulaC6H14O3
InChI KeyXYXCXCJKZRDVPU-NTSWFWBYSA-N
SMILESCCC[C@H](O)[C@H](O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point (clear melt)62.0-71.0?C