Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
CAS: 458-37-7 | C21H20O6 | 368.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR218580100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS555-44-2
IUPAC Name1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Molecular FormulaC51H98O6
InChI KeyPVNIQBQSYATKKL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=98.5 %