Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR218212500
Price (MYR)
942.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
942.00
Add to cart
Chemical Identifiers
CAS75-03-6
IUPAC Nameiodoethane
Molecular FormulaC2H5I
InChI KeyHVTICUPFWKNHNG-UHFFFAOYSA-N
SMILESCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizersilver
Appearance (Color)Clear colorless to light yellow to pink
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
View more
Coupling reagent
RUO – Research Use Only