Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number ACR218210010
Price (MYR)
2,783.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,783.00
Add to cart
Chemical Identifiers
CAS2958-62-5
IUPAC Name2-isocyanato-1,3,5-trimethylbenzene
Molecular FormulaC10H11NO
InChI KeyHJHZRZFONUPQAA-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(N=C=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)42-48?C
Assay (GC)≥97.5%
FormCrystalline or fused solid or lumps or powder or crystalline powder
Appearance (Color)White to pale cream to pale yellow
Coupling reagent
RUO – Research Use Only