Brucine sulfate hydrate, 98+%, Thermo Scientific™
Brucine sulfate hydrate, 98+%, Thermo Scientific™
Brucine sulfate hydrate, 98+%, Thermo Scientific™
Brucine sulfate hydrate, 98+%, Thermo Scientific™
Brucine sulfate hydrate, 98+%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR218170250
Price (MYR)
1.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1.00
Add to cart
Specifications
Chemical Name or MaterialBrucine sulfate hydrate
CAS652154-10-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.