Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR217500500
Price (MYR)
1,653.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,653.00
Add to cart
Chemical Identifiers
CAS16731-55-8
IUPAC Namedipotassium sulfinatosulfonate
Molecular FormulaK2O5S2
InChI KeyRWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95% min