Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals
CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR217500100
Price (MYR)
655.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
655.00
Add to cart
Specifications
Chemical Name or MaterialBis(triphenylphosphine)nickel(II)chloride
Name Note98%
CAS14264-16-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause cancer.