n-Pentane, +99%, extra pure, anhydrous, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, anhydrous, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, anhydrous, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, anhydrous, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number ACR217240025
Price (MYR)
1,278.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,278.00
Add to cart
Specifications
Chemical Name or Materialn-Pentane
Melting Point-130.0°C
Name Notep.a.
CAS109-66-0
CAS Min %99.0