Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR217115000
Price (MYR)
477.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
477.00
Add to cart
Chemical Identifiers
CAS628-21-7
IUPAC Name1,4-diiodobutane
Molecular FormulaC4H8I2
InChI KeyROUYUBHVBIKMQO-UHFFFAOYSA-N
SMILESICCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pink
Refractive Index1.6195-1.6225 @ 20?C
Assay (GC)≥98.5%
StabilizerCopper powder
FormLiquid
Biological buffer
RUO – Research Use Only