Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number ACR217110010
Price (MYR)
595.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
595.00
Add to cart
Chemical Identifiers
CAS68399-81-5
IUPAC Name3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}-2-hydroxypropane-1-sulfonic acid
Molecular FormulaC7H17NO7S
InChI KeyRZQXOGQSPBYUKH-UHFFFAOYNA-N
SMILESOCC(CO)(CO)NCC(O)CS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration with NaOH>=99 %
Water=<1 % (K.F.)
pH4.3 to 5.3 (1 % in water at 25°C)
View more
Biological buffer
RUO – Research Use Only