Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR217085000
Price (MYR)
503.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
503.00
Add to cart
Chemical Identifiers
CAS118994-90-4
IUPAC Name1,3-oxazole-5-carboxylic acid
Molecular FormulaC4H3NO3
InChI KeyQCGMEWVZBGQOFN-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)>98.0 to <102.0%
Assay (HPLC)>98.0%
Chloride content(as NaCl) < 1.0% by titration
Biological buffer, Lewis acid
RUO – Research Use Only