Brij™ 30, Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number ACR216725000
Price (MYR)
701.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
701.00
Add to cart
Chemical Identifiers
CAS89-62-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormPowder
Identification (FTIR)Conforms
Appearance (Color)Orange
Melting Point (clear melt)108.0-118.0°C