4-Acetophenetidide, 97%, Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
CAS: 62-44-2 | C10H13NO2 | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR216591000
Price (MYR)
469.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Chemical Identifiers
CAS58-63-9
IUPAC Name9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H12N4O5
InChI KeyUGQMRVRMYYASKQ-YPLCUDRINA-N
SMILESOC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrophotometry98 to 102 % (lambda max. 248.5nm, pH 7)
Sulfated ash=<0.1 %
Ratio of Absorbances1.65 to 1.77 (A250/A260)
HPLC>=98.5 %
Appearance (Color)White