tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR216221000100 gGlass bottle
Catalog number ACR216221000
Price (MYR)
818.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
818.00
Add to cart
Specifications
Chemical Name or Materialtert-Butyl nitrite
CAS540-80-7
Health Hazard 1GHS Signal Word: Danger
ColorYellow
Density0.8600g/mL