Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
CAS: 1270-98-0 | C5H5Cl3Ti | 219.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number ACR215770025
Price (MYR)
865.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
865.00
Add to cart
Chemical Identifiers
CAS6117-80-2
IUPAC Name(2Z)-but-2-ene-1,4-diol
Molecular FormulaC4H8O2
InChI KeyORTVZLZNOYNASJ-UPHRSURJSA-N
SMILESOC\C=C/CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 % (cis-isomer)
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4770 to 1.4800 (20°C, 589 nm)