Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR215751000
Price (MYR)
444.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Chemical Identifiers
CAS36692-49-6
IUPAC Namemethyl 3,4-diaminobenzoate
Molecular FormulaC8H10N2O2
InChI KeyIOPLHGOSNCJOOO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown to dark brown to dark red
Assay (GC)≥97.5%
Melting Point (clear melt)103.0-110.0?C
FormPowder
Adsorbent
RUO – Research Use Only