Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Thermo Scientific Chemicals
Phenylmethanesulfonyl fluoride, 99%
CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
5 g
10 g
Packaging:
Glass bottle
Catalog number ACR215740500
Price (MYR)
2,742.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,742.00
Add to cart
Chemical Identifiers
CAS429-42-5
IUPAC Nametetrabutylazanium; tetrafluoroboranuide
Molecular FormulaC16H36BF4N
InChI KeyNNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder