Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Thermo Scientific Chemicals
Phenylmethanesulfonyl fluoride, 99%
CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
50 g
Packaging:
Glass bottle
Catalog number ACR215740100
Price (MYR)
1,054.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,054.00
Add to cart
Chemical Identifiers
CAS13472-36-1
IUPAC Nametetrasodium decahydrate (phosphonatooxy)phosphonate
Molecular FormulaH20Na4O17P2
InChI KeyVZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Total nitrogen compounds=<0.001 % (as N)
Potassium (K)=<50 ppm
Impurity=<0.1 % (ortho-phosphate (as PO4))
Appearance (Color)White