Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Thermo Scientific Chemicals
Phenylmethanesulfonyl fluoride, 99%
CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR215740050
Price (MYR)
727.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
727.00
Add to cart
Chemical Identifiers
CAS1605-65-8
IUPAC Namebis(dimethylamino)phosphinoyl chloride
Molecular FormulaC4H12ClN2OP
InChI KeyWYLQARGYFXBZMD-UHFFFAOYSA-N
SMILESCN(C)P(Cl)(=O)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4650 to 1.4680 (20°C, 589 nm)
Titration Argentometric88 to 112 % (ex Cl)
Appearance (Color)Clear colorless to light yellow