(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
CAS: 999-81-5 | C5H13Cl2N | 158.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR215420250
Price (MYR)
519.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
519.00
Add to cart
Chemical Identifiers
CAS14371-10-9
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6210 to 1.6230 (20°C, 589 nm)
Appearance (Color)Clear yellow
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Phase transfer catalyst
RUO – Research Use Only