Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number ACR215370250
Price (MYR)
616.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
616.00
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.005%
Impurity contentCl 0.005%
Impurity contentFe 0.001%
Impurity contentHg 0.1ppm
Impurity contentK 0.02%