Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number ACR215370100
Price (MYR)
452.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Chemical Identifiers
CAS367-57-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3855-1.3900 @ 20?C
Appearance (Color)Clear colorless to yellow
CommentMaterial sourced in UK and US
Assay (GC)≥97.5% (sum of tautomers)