Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR215351000
Price (MYR)
563.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
563.00
Add to cart
Specifications
Chemical Name or MaterialSqualane
Acidity0.2mg KOH/g max.
CAS111-01-3
Melting Point0.3°C