Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR215331000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1159-86-0
IUPAC Name(2-benzoylphenyl)(phenyl)methanone
Molecular FormulaC20H14O2
InChI KeyOJLABXSUFRIXFL-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CC=C1C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystalline powder
Melting Point (clear melt)142-150?C
Appearance (Color)White to yellow