Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR215330025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCitric acid, monosodium salt
Name Note99%
CAS18996-35-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement