Potassium indigotrisulfonate
Potassium indigotrisulfonate
Potassium indigotrisulfonate
Potassium indigotrisulfonate
Thermo Scientific Chemicals
Potassium indigotrisulfonate
CAS: 67627-18-3 | C16H7K3N2O11S3 | 616.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR215170010
Price (MYR)
809.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
809.00
Add to cart
Chemical Identifiers
CAS15525-64-1
IUPAC Namesilver(1+) 2,4-dioxopentan-3-ide
Molecular FormulaC5H7AgO2
InChI KeyUEGHNEVWUGTMES-UHFFFAOYSA-N
SMILES[Ag+].CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%